Assay Details
Gene Symbol: MFAP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015285386.2 | XP_015140872.1 | 3 | 4220 | 96 | |
XM_025150165.1 | XP_025005933.1 | 4 | 4362 | 96 | ||
XM_025150166.1 | XP_025005934.1 | 3 | 5304 | 96 | ||
XM_025150167.1 | XP_025005935.1 | 3 | 4138 | 96 | ||
XM_025150168.1 | XP_025005936.1 | 4 | 4104 | 96 | ||
XM_025150169.1 | XP_025005937.1 | 4 | 4105 | 96 | ||
XM_426287.6 | XP_426287.2 | 3 | 4218 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
