Assay Details
Gene Symbol: TMSB4Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004944973.3 | XP_004945030.1 | 1 - 2 | 63 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004944973.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
