Assay Details
Gene Symbol: PRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004945690.3 | XP_004945747.1 | 3 | 2548 | 74 | |
XM_015279597.1 | XP_015135083.1 | 2 | 2385 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
