Assay Details
Gene Symbol: MYL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015300304.2 | XP_015155790.1 | 3 - 4 | 285 | 78 | |
XM_015300305.2 | XP_015155791.1 | 3 - 4 | 285 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
