Assay Details
Gene Symbol: LOC414835 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015282913.2 | XP_015138399.1 | 1 | 1658 | 79 | |
XM_015282914.2 | XP_015138400.1 | 1 | 1658 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015282913.2, XM_015282914.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
