Assay Details
Gene Symbol: ADRA2BL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025154909.1 | XP_025010677.1 | 1 | 522 | 75 | |
GenBank mRNA | BX931776.2 | - | 1 | 316 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025154909.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
