Assay Details
Gene Symbol: FFAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003641481.4 | XP_003641529.3 | 2 - 3 | 1244 | 114 | |
GenBank mRNA | JQ437358.1 | - | 2 - 3 | 657 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
