Assay Details
Gene Symbol: AES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015299736.2 | XP_015155222.1 | 3 - 4 | 381 | 94 | |
XM_015299737.2 | XP_015155223.1 | 3 - 4 | 385 | 94 | ||
XM_015299738.2 | XP_015155224.1 | 3 - 4 | 362 | 94 | ||
XM_015299739.2 | XP_015155225.1 | 3 - 4 | 360 | 94 | ||
GenBank mRNA | BX933376.2 | - | 3 - 4 | 340 | 94 | |
BX934193.2 | - | 3 - 4 | 265 | 94 | ||
BX934225.1 | - | 3 - 4 | 265 | 94 | ||
CR338750.1 | - | 3 - 4 | 241 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
