Assay Details
Gene Symbol: TP53I3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004935793.3 | XP_004935850.1 | 2 - 3 | 602 | 100 | |
XM_004935795.3 | XP_004935852.1 | 2 - 3 | 296 | 100 | ||
XM_025149105.1 | XP_025004873.1 | 2 - 3 | 455 | 100 | ||
XM_419987.5 | XP_419987.2 | 2 - 3 | 220 | 100 | ||
GenBank mRNA | BX934685.1 | - | 2 - 3 | 603 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
