Assay Details
Gene Symbol: CACNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004937775.3 | XP_004937832.1 | 3 - 4 | 817 | 100 | |
XM_425482.5 | XP_425482.2 | 2 - 3 | 1360 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
