Assay Details
Gene Symbol: LIME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015296605.2 | XP_015152091.1 | 1 - 2 | 329 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015296605.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
