Assay Details
Gene Symbol: LOC101748581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004934965.3 | XP_004935022.1 | 261 | 75 | ||
XM_025143770.1 | XP_024999538.1 | 376 | 75 | |||
XM_025143771.1 | XP_024999539.1 | 293 | 75 | |||
XM_025143772.1 | XP_024999540.1 | 292 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
