Assay Details
Gene Symbol: ARTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291307.2 | XP_015146793.1 | 2 - 3 | 146 | 92 | |
XM_025153107.1 | XP_025008875.1 | 2 - 3 | 206 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
