Assay Details
Gene Symbol: MYO15L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015295329.2 | XP_015150815.1 | 17 - 18 | 2201 | 152 | |
XM_015295330.2 | XP_015150816.1 | 17 - 18 | 2159 | 152 | ||
XM_025141801.1 | XP_024997569.1 | 15 - 16 | 2505 | 152 | ||
XM_025141802.1 | XP_024997570.1 | 15 - 16 | 2502 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
