Assay Details
Gene Symbol: C1QTNF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001234805.5 | XP_001234806.2 | 2 - 3 | 349 | 70 | |
XM_015279159.2 | XP_015134645.1 | 2 - 3 | 388 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
