Assay Details
Gene Symbol: MOB3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004949585.3 | XP_004949642.1 | 2 - 3 | 393 | 94 | |
XM_025144872.1 | XP_025000640.1 | 3 - 4 | 494 | 94 | ||
XM_025144879.1 | XP_025000647.1 | 3 - 4 | 881 | 94 | ||
XM_025144881.1 | XP_025000649.1 | 3 - 4 | 472 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004949585.3, XM_025144872.1, XM_025144879.1, XM_025144881.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
