Assay Details
Gene Symbol: COL2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015279683.2 | XP_015135169.1 | 19 - 20 | 2079 | 115 | |
XM_025156090.1 | XP_025011858.1 | 19 - 20 | 2079 | 115 | ||
XM_025156091.1 | XP_025011859.1 | 19 - 20 | 2079 | 115 | ||
XM_025156092.1 | XP_025011860.1 | 19 - 20 | 2079 | 115 | ||
XM_025156093.1 | XP_025011861.1 | 19 - 20 | 2079 | 115 | ||
XM_025156094.1 | XP_025011862.1 | 19 - 20 | 2079 | 115 | ||
XM_025156095.1 | XP_025011863.1 | 19 - 20 | 2079 | 115 | ||
XM_025156096.1 | XP_025011864.1 | 19 - 20 | 2285 | 115 | ||
XM_025156097.1 | XP_025011865.1 | 15 - 16 | 1936 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
