Assay Details
Gene Symbol: MAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015286091.2 | XP_015141577.1 | 1 - 2 | 141 | 62 | |
XM_015286092.2 | XP_015141578.1 | 1 - 2 | 136 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
