Assay Details
Gene Symbol: P2RY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004938998.3 | XP_004939055.3 | 1 - 2 | 291 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004938998.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
