Assay Details
Gene Symbol: ALDH3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_426371.6 | XP_426371.2 | 2 - 3 | 479 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ALDH3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_426371.6 | XP_426371.2 | 2 - 3 | 479 | 97 |
Set Membership: |
![]() ![]() ![]() |