Assay Details
Gene Symbol: B4GALT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015296479.2 | XP_015151965.1 | 1 - 2 | 257 | 72 | |
XM_025142322.1 | XP_024998090.1 | 1 - 2 | 259 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
