Assay Details
Gene Symbol: MAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025143837.1 | XP_024999605.1 | 133 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025143837.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
