Assay Details
Gene Symbol: IL22RA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001233761.5 | XP_001233762.3 | 3 - 4 | 450 | 70 | |
XM_025149285.1 | XP_025005053.1 | 4 - 5 | 2473 | 70 | ||
XM_025149286.1 | XP_025005054.1 | 3 - 4 | 578 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
