Assay Details
Gene Symbol: AKAP7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025147874.1 | XP_025003642.1 | 1 - 2 | 1100 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AKAP7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025147874.1 | XP_025003642.1 | 1 - 2 | 1100 | 97 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |