Assay Details
Gene Symbol: GPR153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015297062.2 | XP_015152548.1 | 3 - 4 | 1036 | 100 | |
XM_015297063.2 | XP_015152549.1 | 3 - 4 | 1554 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
