Assay Details
Gene Symbol: GPR153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015297062.2 | XP_015152548.1 | 4 - 5 | 1227 | 98 | |
XM_015297063.2 | XP_015152549.1 | 4 - 5 | 1745 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
