Assay Details
Gene Symbol: MUC5ACL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025151100.1 | XP_025006868.1 | 27 - 28 | 17263 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MUC5ACL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025151100.1 | XP_025006868.1 | 27 - 28 | 17263 | 108 |
Set Membership: |
![]() ![]() ![]() |