Assay Details
Gene Symbol: LOC416169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004944797.3 | XP_004944854.1 | 8 - 9 | 2680 | 114 | |
XM_004944798.3 | XP_004944855.1 | 8 - 9 | 2177 | 114 | ||
XM_015293604.2 | XP_015149090.1 | 8 - 9 | 2680 | 114 | ||
XM_015293605.2 | XP_015149091.1 | 8 - 9 | 2680 | 114 | ||
XM_015293607.2 | XP_015149093.1 | 8 - 9 | 2680 | 114 | ||
XM_015293608.2 | XP_015149094.1 | 7 - 8 | 2506 | 114 | ||
XM_015293609.2 | XP_015149095.1 | 8 - 9 | 2284 | 114 | ||
XM_015293610.2 | XP_015149096.1 | 8 - 9 | 2258 | 114 | ||
XM_414501.6 | XP_414501.4 | 8 - 9 | 2578 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
