Assay Details
Gene Symbol: SEOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HM162873.1 | - | 299 | 94 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SEOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HM162873.1 | - | 299 | 94 |