Assay Details
Gene Symbol: RJ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HM771287.1 | - | 3021 | 175 | ||
HM771288.1 | - | 3018 | 175 | |||
HM771289.1 | - | 3021 | 175 | |||
HM771290.1 | - | 3021 | 175 | |||
HM771291.1 | - | 3021 | 175 | |||
HM771292.1 | - | 3021 | 175 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
