Assay Details
Gene Symbol: AKAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004842.3 | NP_004833.1 | 1 - 2 | 459 | 98 | |
GenBank mRNA | AB451273.1 | - | 1 - 2 | 50 | 98 | |
AB451400.1 | - | 1 - 2 | 50 | 98 | ||
AK311820.1 | - | 1 - 2 | 185 | 98 | ||
BC016927.1 | - | 1 - 2 | 137 | 98 | ||
BC073847.1 | - | 1 - 2 | 195 | 98 | ||
BC094732.1 | - | 1 - 2 | 197 | 98 | ||
BC110897.1 | - | 1 - 2 | 151 | 98 | ||
BC118601.1 | - | 1 - 2 | 237 | 98 | ||
BC118606.1 | - | 1 - 2 | 237 | 98 | ||
BC128398.1 | - | 1 - 2 | 237 | 98 | ||
BC128399.1 | - | 1 - 2 | 237 | 98 | ||
BC141926.1 | - | 1 - 2 | 237 | 98 | ||
BC141927.1 | - | 1 - 2 | 237 | 98 | ||
DA144596.1 | - | 1 - 2 | 459 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
