Assay Details
Gene Symbol: NEDD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006156.2 | NP_006147.1 | 1 - 2 | 128 | 95 | |
GenBank mRNA | BC038290.2 | - | 1 - 2 | 61 | 95 | |
BC104200.1 | - | 1 - 2 | 113 | 40031350 | 95 | |
BC104201.1 | - | 1 - 2 | 113 | 40031351 | 95 | |
BC104664.1 | - | 1 - 2 | 129 | 6560920 | 95 | |
BE548741.1 | - | 1 - 2 | 105 | 93 | ||
CF130271.1 | - | 1 - 2 | 128 | 95 | ||
D23662.1 | - | 1 - 2 | 124 | 95 |
Gene Symbol: NEDD8-MDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CB070148.1 | - | 1 - 2 | 96 | 95 | |
DN993851.1 | - | 1 - 2 | 115 | 95 | ||
FY211956.1 | - | 1 - 2 | 43 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC104200.1, BC104201.1, BC104664.1, D23662.1, NM_006156.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
