Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017258.hg.1 |
ENST00000256078.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017258.hg.1 |
ENST00000311936.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017258.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017258.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12017258.hg.1 |
TC12001314.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000556131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000556131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000556131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000556131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000556131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000557334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000557334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000557334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000557334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000557334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000557334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
ENST00000557334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176875.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176875.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176875.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176875.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176875.hg.1 |
TC1200010158.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200176875.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200176875.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176875.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200176875.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000256078.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000311936.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000556131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000556131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000556131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000556131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000556131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000557334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000557334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000557334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000557334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000557334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000557334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
ENST00000557334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_004985.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010158.hg.1 |
NM_033360.3 |
Refseq |