Assay Details
Gene Symbol: RASGEF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282862.1 | NP_001269791.1 | 5 - 6 | 786 | 63 | |
NM_145313.3 | NP_660356.2 | 5 - 6 | 758 | 63 | ||
XM_005271809.3 | XP_005271866.1 | 4 - 5 | 636 | 63 | ||
XM_011539500.2 | XP_011537802.1 | 4 - 5 | 746 | 63 | ||
GenBank mRNA | AK095136.1 | - | 5 - 6 | 786 | 63 | |
AK127432.1 | - | 5 - 6 | 3184 | 63 | ||
BC022548.1 | - | 5 - 6 | 758 | 4814750 | 63 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001200.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10015057.hg.1 | ENST00000374459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10015057.hg.1 | ENST00000395809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10015057.hg.1 | ENST00000395810.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10015057.hg.1 | ENST00000472864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10015057.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10015057.hg.1 | TC10001200.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374455.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374455.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374455.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374455.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374455.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374455.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000472864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000472864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000472864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000472864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | ENST00000472864.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374455.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374455.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374455.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374455.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374455.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374455.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000472864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000472864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000472864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000472864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | ENST00000472864.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010432.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | ENST00000374455.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | ENST00000472864.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | NM_145313.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | TC1000010432.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | ENST00000374459.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | ENST00000374455.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | ENST00000395809.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | ENST00000395810.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | ENST00000472864.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | NM_001282862.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000170939.hg.1 | NM_145313.3 | Refseq |
