Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003769.hg.1 |
ENST00000325239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003769.hg.1 |
ENST00000413659.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003769.hg.1 |
ENST00000426033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003769.hg.1 |
ENST00000465910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003769.hg.1 |
ENST00000544136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003769.hg.1 |
NM_020945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003769.hg.1 |
TC10000323.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003768.hg.1 |
ENST00000325239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003768.hg.1 |
ENST00000413659.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003768.hg.1 |
ENST00000426033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003768.hg.1 |
ENST00000465910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003768.hg.1 |
ENST00000544136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003768.hg.1 |
NM_020945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10003768.hg.1 |
TC10000323.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000151601.hg.1 |
ENST00000265453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151601.hg.1 |
ENST00000325239.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151601.hg.1 |
ENST00000465910.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151601.hg.1 |
NM_020945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000151601.hg.1 |
TC1000007574.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000151601.hg.1 |
NM_020945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000151601.hg.1 |
ENST00000265453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151601.hg.1 |
ENST00000325239.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151601.hg.1 |
ENST00000465910.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151600.hg.1 |
ENST00000265453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151600.hg.1 |
ENST00000325239.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151600.hg.1 |
ENST00000465910.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151600.hg.1 |
NM_020945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000151600.hg.1 |
TC1000007574.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000151600.hg.1 |
ENST00000265453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151600.hg.1 |
ENST00000325239.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151600.hg.1 |
ENST00000465910.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000151600.hg.1 |
NM_020945.1 |
Refseq |