Assay Details
Gene Symbol: RIDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005836.2 | NP_005827.1 | 3 - 4 | 323 | 125 | |
GenBank mRNA | BC010280.1 | - | 3 - 4 | 291 | 3048297 | 125 |
BC012592.1 | - | 3 - 4 | 324 | 4102470 | 125 | |
BC093059.1 | - | 3 - 4 | 304 | 30370716 | 125 | |
CR456844.1 | - | 3 - 4 | 229 | 125 | ||
CR541652.1 | - | 3 - 4 | 229 | 125 | ||
X95384.1 | - | 3 - 4 | 323 | 125 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001461.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
kidney development
regulation of translational termination
brain development
negative regulation of translation
lung development
response to lipid
negative regulation of epithelial cell proliferation
G1 to G0 transition
RNA phosphodiester bond hydrolysis, endonucleolytic
response to salt
RNA binding
endoribonuclease activity, producing 3'-phosphomonoesters
long-chain fatty acid binding
protein homodimerization activity
transition metal ion binding
kidney development
regulation of translational termination
brain development
negative regulation of translation
lung development
response to lipid
negative regulation of epithelial cell proliferation
G1 to G0 transition
RNA phosphodiester bond hydrolysis, endonucleolytic
response to salt
RNA binding
endoribonuclease activity, producing 3'-phosphomonoesters
long-chain fatty acid binding
protein homodimerization activity
transition metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000519155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000519155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08018905.hg.1 | ENST00000254878.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08018905.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08018905.hg.1 | TC08001461.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08018906.hg.1 | ENST00000254878.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08018906.hg.1 | ENST00000520989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08018906.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08018906.hg.1 | TC08001461.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000519155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000519155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011208.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176266.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176266.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176266.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176266.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176266.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176266.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176266.hg.1 | TC0800011208.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800176266.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176266.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176266.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176266.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176266.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176266.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176267.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176267.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176267.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176267.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176267.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176267.hg.1 | NM_005836.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800176267.hg.1 | TC0800011208.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800176267.hg.1 | ENST00000254878.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176267.hg.1 | ENST00000519608.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176267.hg.1 | ENST00000520507.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176267.hg.1 | ENST00000521560.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176267.hg.1 | ENST00000522791.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800176267.hg.1 | NM_005836.2 | Refseq |
