Assay Details
Gene Symbol: SEC22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF100749.1 | - | 1 - 2 | 45 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF100749.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
