Assay Details
Gene Symbol: MYCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033082.2 | NP_001028254.2 | 1 - 2 | 207 | 116 | |
NM_005376.4 | NP_005367.2 | 1 - 2 | 207 | 116 | ||
GenBank mRNA | BC011864.2 | - | 1 - 2 | 95 | 4541675 | 116 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002551.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC011864.2, NM_001033082.2, NM_005376.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000450953.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | ENST00000450953.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013811.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039682.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039682.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039682.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039682.hg.1 | ENST00000429311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039682.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039682.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039682.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039682.hg.1 | TC01002551.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008065.hg.1 | ENST00000372451.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039680.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039680.hg.1 | ENST00000429311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039680.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039680.hg.1 | TC01002551.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01039687.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039687.hg.1 | ENST00000429311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039687.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039687.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039687.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039687.hg.1 | TC01002551.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01039684.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039684.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039684.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039684.hg.1 | ENST00000429311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039684.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039684.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039684.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039684.hg.1 | TC01002551.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01039681.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039681.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039681.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039681.hg.1 | ENST00000429311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039681.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039681.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039681.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039681.hg.1 | TC01002551.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0100203326.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203326.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203326.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203326.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203326.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203326.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203326.hg.1 | TC0100013811.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100203326.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203326.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203326.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203326.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203326.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203326.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203321.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203321.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203321.hg.1 | TC0100013811.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100203321.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203321.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008065.hg.1 | ENST00000372451.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | ENST00000450953.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | TC0100013811.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | ENST00000450953.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203323.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203330.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203330.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203330.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203330.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203330.hg.1 | TC0100013811.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100203330.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203330.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203330.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203330.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203320.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203320.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100203320.hg.1 | TC0100013811.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100203320.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100203320.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000372815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000372816.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000397332.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000450953.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | ENST00000450953.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033081.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_001033082.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_005376.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013811.hg.1 | NM_005376.4 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 163352 | - | - |
