Assay Details
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085399.1 | NP_001078868.1 | 1 - 2 | 180 | 73 | |
NM_001085400.1 | NP_001078869.1 | 1 - 2 | 180 | 73 | ||
GenBank mRNA | BC039540.1 | - | 1 - 2 | 178 | 5748847 | 73 |
DA400382.1 | - | 1 - 2 | 180 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001116.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04015239.hg.1 | ENST00000314117.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015239.hg.1 | ENST00000454158.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015239.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04015239.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04015239.hg.1 | TC04001116.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04015241.hg.1 | ENST00000314117.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015241.hg.1 | ENST00000454158.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015241.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04015241.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04015241.hg.1 | TC04001116.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000512114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000512114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000512114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000512114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | ENST00000512114.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171070.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171070.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171070.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171070.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171070.hg.1 | TC0400010418.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400171070.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171070.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171070.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171070.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000512114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000512114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000512114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000512114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | ENST00000512114.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010418.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171069.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171069.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171069.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171069.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171069.hg.1 | TC0400010418.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400171069.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171069.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171069.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171069.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171067.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171067.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171067.hg.1 | ENST00000512114.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171067.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171067.hg.1 | NM_001085400.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171067.hg.1 | TC0400010418.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400171067.hg.1 | ENST00000314117.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171067.hg.1 | ENST00000454158.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171067.hg.1 | ENST00000512114.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171067.hg.1 | NM_001085399.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171067.hg.1 | NM_001085400.1 | Refseq |
