Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034838.hg.1 |
ENST00000237696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034838.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034838.hg.1 |
TC03001949.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000462663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000462663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000462663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000471444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000471444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000479756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000479756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000479756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000479756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000479756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000479756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000498640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000498640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000498640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
ENST00000498640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_002888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_002888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_002888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_002888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_002888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_002888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_002888.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196728.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196728.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196728.hg.1 |
TC0300012945.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300196728.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196728.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196729.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196729.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196729.hg.1 |
TC0300012945.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300196729.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196729.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000237696.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000462663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000462663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000462663.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000471444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000471444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000479756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000479756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000479756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000479756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000479756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000479756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000498640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000498640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000498640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
ENST00000498640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_002888.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_002888.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_002888.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_002888.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_002888.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_002888.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_002888.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012945.hg.1 |
NM_206963.1 |
Refseq |