Assay Details
Gene Symbol: GRIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006718823.1 | XP_006718886.1 | 14 - 15 | 2418 | 66 | |
XM_011542775.2 | XP_011541077.1 | 14 - 15 | 2788 | 66 | ||
XM_011542776.2 | XP_011541078.1 | 12 - 13 | 4322 | 66 | ||
XM_011542777.2 | XP_011541079.1 | 11 - 12 | 4192 | 66 | ||
XM_017017611.1 | XP_016873100.1 | 11 - 12 | 2429 | 66 | ||
GenBank mRNA | U16129.1 | - | 13 - 14 | 2298 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
