Assay Details
Gene Symbol: CYP39A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278738.1 | NP_001265667.1 | 10 - 11 | 1440 | 104 | |
NM_001278739.1 | NP_001265668.1 | 9 - 10 | 1325 | 104 | ||
NM_016593.4 | NP_057677.2 | 10 - 11 | 1500 | 104 | ||
XM_017010924.1 | XP_016866413.1 | 9 - 10 | 1367 | 104 | ||
GenBank mRNA | AF237982.1 | - | 10 - 11 | 1296 | 104 | |
AK292263.1 | - | 10 - 11 | 1432 | 104 | ||
AK301604.1 | - | 10 - 11 | 1318 | 104 | ||
AK307986.1 | - | 9 - 10 | 989 | 104 | ||
BC010358.1 | - | 10 - 11 | 1460 | 4334109 | 104 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001772.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06021325.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06021325.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06021325.hg.1 | TC06001772.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000476076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000476076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000489657.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000489657.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000476076.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000476076.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000480804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000489657.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000489657.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011948.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600182240.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600182240.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600182240.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600182240.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600182240.hg.1 | NM_016593.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600182240.hg.1 | TC0600011948.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600182240.hg.1 | NM_001278739.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600182240.hg.1 | ENST00000275016.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600182240.hg.1 | ENST00000619708.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600182240.hg.1 | NM_001278738.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600182240.hg.1 | NM_016593.4 | Refseq |
