Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018371.hg.1 |
TC19001292.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018371.hg.1 |
ENST00000392386.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018371.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000593286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000593286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000594325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000594325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000594325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000594325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000596360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000596360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000596360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000597131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000597131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000597131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
ENST00000597131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000593286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000593286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000594325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000594325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000594325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000594325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000596360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000596360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000596360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000597131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000597131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000597131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
ENST00000597131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010025.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182408.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182408.hg.1 |
ENST00000597131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182408.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182408.hg.1 |
TC1900010025.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900182408.hg.1 |
NM_004750.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182408.hg.1 |
ENST00000392386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182408.hg.1 |
ENST00000597131.1 |
Ensembl |