Assay Details
Gene Symbol: ZBTB11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024407.1 | 1 | 1955 | 97 | ||
GenBank mRNA | AK022880.1 | - | 1 | 1939 | 97 | |
BC016930.1 | - | 1 | 1910 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger and BTB domain containing 11 | ZBTB11 |
ZNF-U69274, ZNF913 |
-396.0 | Sense | Chr. 3 - 101649439 - 101677144 on NCBI Build GRCh38 |
Downstream | ribosomal protein L24 | RPL24 |
HEL-S-310, L24 |
2.7kb | Sense | Chr. 3 - 101681090 - 101686719 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000522.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 539876 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P306479 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P33561 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P33561 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 40724 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 16080 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 30158 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 52403 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P33561 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P306479 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P306479 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_32_P33561 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591954 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591963 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591967 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591977 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591972 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22592005 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591942 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15469639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591943 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453666 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453668 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591958 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591978 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591976 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591947 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591957 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453706 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591961 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453690 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591968 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591953 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591949 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591948 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591970 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591940 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591945 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591974 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591936 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591973 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591968 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591953 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591958 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591945 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591936 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591976 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591967 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591970 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591974 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453706 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591977 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591940 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591961 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22592005 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591943 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591963 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453666 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591978 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591942 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15469639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591949 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591973 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591957 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591948 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591954 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591972 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453668 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02453690 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22591947 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P33561 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00771364 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P306479 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P33561 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00771364 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P33567 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453703 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15469623 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453691 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453695 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453765 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453699 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453689 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453685 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453673 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453677 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453693 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15469628 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453768 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453702 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453664 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453687 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453669 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453680 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453666 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15469627 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15469630 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453667 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453671 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453686 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453683 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453764 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453697 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453770 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02453706 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00771364 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8081359 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8081358 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 232259_PM_s_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 232260_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR03009696.hg.1 | ENST00000536865.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03009696.hg.1 | NR_024407.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03009696.hg.1 | TC03000522.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03009697.hg.1 | ENST00000536865.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03009697.hg.1 | NR_024407.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03009697.hg.1 | TC03000522.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03009698.hg.1 | NR_024407.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03009698.hg.1 | TC03000522.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0300158236.hg.1 | ENST00000536865.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300158236.hg.1 | NR_024407.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300158236.hg.1 | TC0300008176.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300158236.hg.1 | NR_024407.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300158236.hg.1 | ENST00000536865.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300158237.hg.1 | ENST00000536865.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300158237.hg.1 | ENST00000609682.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300158237.hg.1 | NR_024407.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300158237.hg.1 | TC0300008176.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300158237.hg.1 | ENST00000536865.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300158237.hg.1 | ENST00000609682.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300158237.hg.1 | NR_024407.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300158238.hg.1 | ENST00000609682.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300158238.hg.1 | NR_024407.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300158238.hg.1 | TC0300008176.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300158238.hg.1 | ENST00000609682.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300158238.hg.1 | NR_024407.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 232259_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 232260_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 232260_at | - | - |
Human Genome U133 Plus 2.0 Array | 232259_PM_s_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232260_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232260_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232260_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232260_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232259_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232259_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232259_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232259_s_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232259_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232259_s_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232260_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232259_s_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232260_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232260_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232259_s_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232259_s_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232259_s_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232260_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232260_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232259_s_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232260_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232260_at5 | - | - |
Human Genome U95 Set | 80822_PM_at | - | - |
Human Genome U95 Set | 80822_at | - | - |
Human Genome U95 Set | 82871_PM_at | - | - |
Human Genome U95 Set | 82871_at | - | - |
Human Genome U133 Set | 232259_s_at | - | - |
Human Genome U133 Set | 232260_PM_at | - | - |
Human Genome U133 Set | 232260_at | - | - |
Human Genome U133 Set | 232259_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022937 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022937 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 232635 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3237156 | BC016930.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3237156 | CR598311.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3237156 | NR_024407.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3237156 | BC016930.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3237156 | CR598311.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3237156 | NR_024407.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237156 | BC016930.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237156 | CR598311.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237156 | NR_024407.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237156 | BC016930.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237156 | CR598311.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237156 | NR_024407.1 | human_transcriptome |
