Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024578.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024578.hg.1 |
NM_000590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024578.hg.1 |
TC05001807.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012120.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012120.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012120.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012120.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012120.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012120.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012120.hg.1 |
NM_000590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012120.hg.1 |
NM_000590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012120.hg.1 |
NM_000590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012120.hg.1 |
NM_000590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012120.hg.1 |
NM_000590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012120.hg.1 |
NM_000590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183860.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183860.hg.1 |
NM_000590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183860.hg.1 |
TC0500012120.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500183860.hg.1 |
NM_000590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183860.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012120.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012120.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012120.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012120.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012120.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012120.hg.1 |
ENST00000274520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012120.hg.1 |
NM_000590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012120.hg.1 |
NM_000590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012120.hg.1 |
NM_000590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012120.hg.1 |
NM_000590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012120.hg.1 |
NM_000590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012120.hg.1 |
NM_000590.1 |
Refseq |