Assay Details
Gene Symbol: UGCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003358.2 | NP_003349.1 | 4 - 5 | 890 | 125 | |
XM_005252186.4 | XP_005252243.1 | 5 - 6 | 913 | 125 | ||
XM_017015107.1 | XP_016870596.1 | 4 - 5 | 549 | 125 | ||
GenBank mRNA | AK314847.1 | - | 4 - 5 | 495 | 125 | |
BC038711.1 | - | 4 - 5 | 891 | 5295561 | 125 | |
D50840.1 | - | 4 - 5 | 730 | 125 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000560.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09006254.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09006254.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09006254.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09006254.hg.1 | TC09000560.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900156431.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900156431.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900156431.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900156431.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900156431.hg.1 | TC0900008441.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900156431.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900156431.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900156431.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900156431.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900156432.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900156432.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900156432.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900156432.hg.1 | TC0900008441.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900156432.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900156432.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900156432.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000374279.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000474306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000489355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000490110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | ENST00000495085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008441.hg.1 | NM_003358.2 | Refseq |
