Assay Details
Gene Symbol: BOD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148894.2 | NP_683692.2 | 20 - 21 | 8788 | 95 | |
XM_005248150.3 | XP_005248207.1 | 20 - 21 | 8805 | 95 | ||
XM_011513827.2 | XP_011512129.1 | 20 - 21 | 8805 | 95 | ||
XM_011513829.2 | XP_011512131.1 | 20 - 21 | 8805 | 95 | ||
XM_011513830.2 | XP_011512132.1 | 18 - 19 | 8188 | 95 | ||
GenBank mRNA | AB037748.2 | - | 11 - 12 | 4932 | 95 | |
AK025361.1 | - | 6 - 7 | 449 | 95 | ||
BC016987.1 | - | 10 - 11 | 728 | 95 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001047.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04014324.hg.1 | ENST00000040738.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04014324.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04014324.hg.1 | TC04001047.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000509897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000509897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000509897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400169115.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400169115.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400169115.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400169115.hg.1 | TC0400010085.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400169115.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400169115.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400169115.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000509897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000509897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000509897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
