Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012751.hg.1 |
TC22000723.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012751.hg.1 |
ENST00000337843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012751.hg.1 |
ENST00000397110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012751.hg.1 |
ENST00000470655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012751.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012751.hg.1 |
NM_182486.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012749.hg.1 |
TC22000723.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012749.hg.1 |
ENST00000337843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012749.hg.1 |
ENST00000397110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012749.hg.1 |
ENST00000470655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012749.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012749.hg.1 |
NM_182486.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000337843.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000337843.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000337843.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000337843.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000337843.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000337843.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000397110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000397110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000397110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000397110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000397110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000434784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000434784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000434784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000434784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000493023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000493023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000493023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000493023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
ENST00000493023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
NM_182486.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
NM_182486.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
NM_182486.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
NM_182486.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009345.hg.1 |
NM_182486.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200170311.hg.1 |
ENST00000337843.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170311.hg.1 |
ENST00000397110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170311.hg.1 |
ENST00000493023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170311.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200170311.hg.1 |
NM_182486.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200170311.hg.1 |
TC2200009345.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200170311.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200170311.hg.1 |
ENST00000337843.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170311.hg.1 |
ENST00000397110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170311.hg.1 |
ENST00000493023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170311.hg.1 |
NM_182486.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000337843.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000337843.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000337843.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000337843.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000337843.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000337843.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000397110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000397110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000397110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000397110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000397110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000434784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000434784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000434784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000434784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000493023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000493023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000493023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000493023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
ENST00000493023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
NM_031910.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
NM_182486.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
NM_182486.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
NM_182486.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
NM_182486.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009345.hg.1 |
NM_182486.1 |
Refseq |