Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000481127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000481127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000481127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
ENST00000481127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016135.hg.1 |
ENST00000369503.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016135.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016135.hg.1 |
TC01001014.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000481127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000481127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000481127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
ENST00000481127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009534.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168608.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168608.hg.1 |
NM_018420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168608.hg.1 |
TC0100009534.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100168608.hg.1 |
ENST00000369503.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168608.hg.1 |
NM_018420.2 |
Refseq |