Assay Details
Gene Symbol: NRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005261063.3 | XP_005261120.1 | 2 - 3 | 189 | 127 | |
GenBank mRNA | BE792046.1 | - | 2 - 3 | 261 | 127 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005261063.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
ovarian follicle rupture
transcription, DNA-templated
circadian rhythm
lipid storage
androgen receptor signaling pathway
circadian regulation of gene expression
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
cellular response to estradiol stimulus
core promoter sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
estrogen receptor binding
glucocorticoid receptor binding
histone deacetylase binding
retinoid X receptor binding
androgen receptor binding
negative regulation of transcription by RNA polymerase II
ovarian follicle rupture
transcription, DNA-templated
circadian rhythm
lipid storage
androgen receptor signaling pathway
circadian regulation of gene expression
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
cellular response to estradiol stimulus
core promoter sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
estrogen receptor binding
glucocorticoid receptor binding
histone deacetylase binding
retinoid X receptor binding
androgen receptor binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000411932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000411932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000411932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000411932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | ENST00000411932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000318948.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000400202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000411932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000411932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000411932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000411932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | ENST00000411932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | NM_003489.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008545.hg.1 | NM_003489.3 | Refseq |
